3M™ P100 Particulate Filters Edge
Edge
For oil- and non-oil applications
Manufacturer:  3M™ 7000051991
 View more versions of this product
Catalog No. 18-999-2573
Edge
Requires : If used with 3M Respirator 5000 Series or as a prefilter for cartridges on 3M Half- and Full-Facepiece Respirators 6000, 7000 or FF-400 filters require filter adapter, Cat. No. 17-459-4.
Description
Description
Designed for use with 3M 5000 Series Respirators or 6000 Series cartridges with 3M Adapter 502, or 6000, 7000 or FF-400 Series facepieces with bayonet filter holders
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
  • 3M Catalog# 2091 , 3M Catalog# 2096 , 3M Catalog# 2097
Compliance
NIOSH approved under 42 CFR 84 standards for environments containing certain oil and non-oil based particulates (with 99.97% filter efficiency).
Specifications
Specifications
NIOSH, OSHA
4.3 x 7 x 4.3 in. (10.9 x 17.7 x 10.9 cm)
4.3 in.
4.3 in.
7 in.
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, sanding
Particulate Filter
Magenta
2091
10.9 cm
10.9 cm
17.7 cm
3M™ respirator 5000 series or 3M™ cartridge 6000 series with 3M™ adapter 502, or 3M™ half and full facepiece 6000, 7000 and ff-400 series with bayonet filter holders