3M™ Nuvo™ Readers Eyewear
Bifocal readers with three magnification diopters
Manufacturer:  3M™ 114360000020
 View more versions of this product
Catalog No. 19-123-504
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
Designed for workers who have difficulty reading small print or who are engaged in fine detail work.
  • Classic look of a dual lens but with tough, lightweight one-piece lens
  • Lens with anti-fog coating absorbs 99.9% UV
  • Gray, adjustable length temples with soft tips for comfortable fit
  • Soft nosepiece
  • Integral sideshields provide profile protection
  • Cushioned brow bar
  • 3M Catalog# 11434-00000-20, 11435-00000-20, 11436-00000-20, 11500-00000-20, 11501-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Specifications
ANSI Z87.1-2015
+2.50
Scratch-resistant; Anti-fog
Anti-fog,Scratch-resistant
0.999
Designed for workers who have difficulty reading small print or who are engaged in fine detail work
11436-00000-20
Clear
Bifocal Readers