Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
- Ultra-soft foam formulation for all-day comfort
- Tapered shape for easy insertion and canal-hugging seal
- Four vibrant colors in each box: orange, red, pink, and purple
- 3M Catalog# P1000 , 3M Catalog# P1001
Compliance
CSA Class AL.
Specifications
CSA Class AL | |
Vinyl | |
Polyurethane | |
Corded | |
Tapered | |
Regular |
Multi-color | |
For easy insertion and canal-hugging seal | |
P1001 | |
32 dB | |
Nitro™ Earplug |