3M™ Maxim™ Eyewear
Contemporary, athletic styling
Manufacturer:  3M™ 132510000020
 View more versions of this product
Catalog No. 29-635-2690
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
  • 3M Catalog# 13250-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Specifications
Meets ANSI Z87.1-2010 and CSA Z94.3-2007 standards.
Bronze
Adjustable
Anti-fog
Black
Polycarbonate