missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ High-Capacity Chemical Sorbents
Made from inert polypropylene and polyester fibers
Manufacturer: 3M™ 7000002024
Description
- Suitable for a wide range of liquids
- Lightweight, easy to deploy and dust free
- Identifiable coloring for easy identification and visibility
- Can be wrung out and reused
- High absorption capacity minimizes amount of waste for disposal
- L x W: 50 ft. x 5 in. (15.2m x 12.7cm)
- Sorbing Capacity: 31.5 gal. (119.2L)
- 3M Catalog# CFL550DD , 3M Catalog# CRL15150 , 3M Catalog# PD914
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
CFL550DD | |
5 in. | |
OSHA 29 CFR 1910.132 | |
10.5 gal. | |
15.24m | |
High Capacity Chemical Sorbent | |
10.5 gal., 39.74L |
Polypropylene, Polyester | |
12.7cm | |
Yellow | |
39.74L | |
50 ft. | |
For use on most aggressive (hazardous) and non-aggressive liquids | |
50 ft. x 5 in. (15.24m x 12.7cm) |