missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ High-Capacity Chemical Sorbents

Made from inert polypropylene and polyester fibers

Manufacturer:  3M™ 7000001947

 View more versions of this product

Catalog No. 19-083193




Description

Description

  • Suitable for a wide range of liquids
  • Lightweight, easy to deploy and dust free
  • Identifiable coloring for easy identification and visibility
  • Can be wrung out and reused
  • High absorption capacity minimizes amount of waste for disposal
  • L x W: 50 ft. x 5 in. (15.2m x 12.7cm)
  • Sorbing Capacity: 31.5 gal. (119.2L)
  • 3M Catalog# CFL550DD , 3M Catalog# CRL15150 , 3M Catalog# PD914

These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

Specifications

Specifications

OSHA 29 CFR 1910.132
22.98 x 36 cm (9.05 x 14.17 in.)
PD914
88.95 L
22.98 cm
Chemical Sorbent Pad
36 cm
Yellow
For control of spills of certain hazardous liquids
23.5 gal.
9.05 in.
Polyester
14.17 in.
SDS
Documents
Product Certifications