3M™ H-Series White Hood Assemblies
Used on 3M™ Belt-Mounted Powered Air Purifying Respirator (PAPR)s and Supplied Air Respirator Systems
$133.61 - $175.11
Specifications
Product Type Hood Assembly
Material Tychem™ QC Fabric
Size Standard
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
29-663-298 3M™
H421
Inner shroud and cap suspension Each for $133.61
19-0881269 3M™
H412
Collar and hardhat Each for $175.11
19-0881271 3M™
H422*
Inner shroud and hardhat Each for $173.40
Specifications
Description
  • Feature lightweight Tychem™ QC Fabric
  • Comfortable headgear
  • Can be worn with facial hair and glasses
  • White
3M H-400 Series Hoods
  • Feature lightweight Tychem™ QC Fabric and provide high levels of airflow for maximum comfort For pharmaceutical, laboratory, and health care applications as well as during painting and sanding operations H-400 is available with hardhat and choice of inner shroud or collar
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Description & Specifications
Specifications
Hood Assembly
Tychem™ QC Fabric
Standard