3M™ H-Series Hood Assemblies Replacement Hoods
Provide head and neck protection from pesticide and chemical overspray. H-Series hoods are for use with belt-mounted PAPRs or supplied air components.
Manufacturer:  3M™ H41010
Catalog No. 29-663-306
Description
Description
  • Lightweight, loose fitting, hood
  • Replacement for 3M™ Hood Assemblies H-411 and H-412
Specifications
Specifications
Assembly, machine operation
Hood
Collar
Belt mounted powered air purifying respirator or supplied air component
White
Standard
Documents