3M™ Empore™ Vacuum Manifold Accessories
$310.69 - $310.69
Specifications
For Use With (Equipment) EmPore™ vacuum manifold assembly
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity  
14-378-62 3M™
E660
For sealing unused portions of a 96-well solid Phase extraction plate during vacuum processing to enable vacuum to be uniformly applied Sealing tape pad Pack of 10 Each for $310.69
Case of 6 Pack for $1,864.14
Specifications Description & Specifications
Specifications
EmPore™ vacuum manifold assembly