3M™ Empore™ SDB-XC Extraction Disks
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
$691.09 - $4,170.54
Specifications
Cartridge Material Silica
Certifications/Compliance EPA 515.2
Column Phase Reversed
For Use With (Application) Solid Phase extraction
Sorbent SDB-XC
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
14-386-4 3M™
E224047
4.7cm Pack of 20 Each for $691.09
Case of 3 Pack for $2,073.27
14-386-5 3M™
E234090
9cm Case of 30 Each for $4,170.54
Specifications
Description
  • Approved for use in EPA Method 515.2
  • Provide the next step in environmental sample preparation, soluble polar compound extraction; ideal for phenol compounds having poor recoveries by almost any known extraction technique
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes
Description & Specifications
Specifications
Silica
Reversed
SDB-XC
EPA 515.2
Solid Phase extraction