missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ Empore™ SDB-RPS Extraction Disks

Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples

$896.74 - $1,859.34

Specifications

Column Phase Reversed
Solvent Reduction 90%
Sorbent SDB-RPS
For Use With (Application) Analytical preparation of aqueous environmental samples
Certifications/Compliance EPA 515.2
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
14-386-9 3M™
E234190
9cm Pack of 10 Each for $1,859.34
Case of 3 Pack for $5,578.02
14-386-8 3M™
E224147
4.7cm Pack of 20 Each for $896.74
Case of 3 Pack for $2,690.22
Specifications

Description

  • Approved for use in EPA Method 515.2
  • Used to collect the more water-soluble analytes from water samples
  • Adsorbent is effective for explosives, pesticides, phenolics, and drugs of abuse Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes

Description & Specifications

Specifications

Reversed
SDB-RPS
EPA 515.2
90%
Analytical preparation of aqueous environmental samples