3M™ Empore™ Oil and Grease Extraction Disks
Reduce extraction times to an average of eight minutes/liter for most sample matrices
Specifications
For Use With (Application) For use in EPA Method 1664
Includes Hydrophobic matrix
Sorbent C18
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
14-638-30A 3M™
E227047
4.7cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-638-30B 3M™
2370
9cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
  • Designed for use in EPA Method 1664
  • Require almost half the solvent of traditional methods and do not form emulsions during sample preparation
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes
Description & Specifications
Specifications
For use in EPA Method 1664
Hydrophobic matrix
C18