3M™ Empore™ Carbon Extraction Disks
Provides all the advantages of activated carbon in a compact and thin disk
$2,020.03 - $2,020.03
Specifications
For Use With (Application) Extraction of volatile, highly Polar, and water soluble compounds from aqueous solutions
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
14-378-062 3M™
E2272
4.7cm Case of 60 Each for $2,020.03
Specifications
Description
  • Designed for the extraction of volatile, highly polar, and water soluble compounds from aqueous solutions
Description & Specifications
Specifications
Extraction of volatile, highly Polar, and water soluble compounds from aqueous solutions