3M™ Empore™ Anion and Cation Exchange-SR Extraction Disks
Provide significantly faster extraction time with no negative effect on recoveries
$665.06 - $665.06
Specifications
Diameter (Metric) Outer 4.7cm
For Use With (Application) Analytical preparation of aqueous environmental samples
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Solvent Reduction Price Quantity  
Catalog Number Mfr. No. Solvent Reduction Price Quantity  
14-378-104 3M™
E2251
90% CatIon Exchange N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-386-6 3M™
E225247
90% AnIon Exchange Pack of 20 Each for $665.06
Case of 3 Pack for $1,995.18
Specifications
Description
  • Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes
  • Anion exchange disks are approved for use in EPA Methods 549.1 and 552.1
  • Replace liquid/solid procedures for the determination of haloacetic acids in water, as well as chlorinated herbicide Dalapon™
Description & Specifications
Specifications
4.7cm
Analytical preparation of aqueous environmental samples