3M™ Empore™ 96-Well Extraction Disk Plates
High-performance extraction disk plates for high-throughput SPE
$906.39 - $906.39
Catalog Number | Mfr. No. | Sorbent | Volume (Metric) Well | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sorbent | Volume (Metric) Well | Price | Quantity | ||||
14-378-58
|
3M™
E6030SD |
MPC-SD | 2.5 mL |
Each for $906.39
Pack of 12 Each for $10,876.68 |
|
Description
- Process 96 samples in 8 x 12 plate formats compatible with liquid handling technologies and injection systems; samples can be extracted in approximately one hour or less
- Wells contain patented Empore standard-density particle-loaded membrane for efficient sample extraction followed by elution in 100μL with mobile phase or mobile-phase compatible solutions Ideal for sample preparation prior to LC/MS and other high-throughput applications
- C2, C8, or C18 silica sorbents for nonpolar interactions with analytes from complex samples
- Universal resin (UR) sorbent plates contain polar functioning groups to promote a much more generic approach with high retention of acidic, neutral, and base analytes
- Use with 3M Empore Vacuum Manifold Assembly when extracting multiple samples
- Available with 1.2 or 2.5mL reservoir volumes with a 5.5μm disk diameter
Specifications
Universal |