3M™ Empore™ 7mm dia./3mL vol. Extraction Disk Cartridges
Provide a unique alternation to packed column technology
$846.65 - $1,359.34
Specifications
Material Silica
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Resin Composition Sorbent Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Resin Composition Sorbent Price Quantity  
14-388-13 3M™
4215SD
For sample volumes <3mL, Strongly non-Polar analytes Polypropylene C8-SD Case of 150 Each for $1,359.34
36-100-4504 3M™
4245SD
Solid Phase extraction of analytes from complex sample matrices Universal UR-SD Pack of 150 Each for $846.65
Specifications
Description
  • Elution volumes of approximately 0.25mL
  • For sample volumes <3mL
  • Membranes efficiently extract samples with no channeling
  • Small bed volume allows compound elution in 100 to 300μL of mobile-phase or mobile-phase-compatible solutions
  • Efficient extraction followed by direct injection of the solutions
Choice of Two Densities
  • Standard-density (SD) membranes for silica-based sorbent chemistries allow reliable sample preparation for difficult sample matrices
  • High-density (HD) membrane formulation is convenient when optimal extraction efficiency is required
Description & Specifications
Specifications
Silica