3M™ E•A•R™ UltraFit Plus™ Ear Plugs
NRR: 26dB
$127.93 - $188.14
Specifications
Certifications/Compliance CSA Class AL
Color Yellow
Corded Corded
Product Type Ultrafit Plus™ Earplug
Size Universal
View More Specs
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Products 2
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
29-635-123
View Documents
3M™
3406004
Without case
Pack of 100 Pair for $188.14
Case of 4 Pack for $711.88
29-635-121
View Documents
3M™
3406002
Case
Pack of 50 Pair for $127.93
Case of 4 Pack for $484.08
Description
Description
  • Colorful patented grips make insertion easy
  • No rolling or touching during insertion
  • Washable—promotes worker hygiene
  • Patented flange design for most comfortable fit of any reusable plug
  • Corded
  • 3M Catalog# 340-6002 , 3M Catalog# 340-6004
Specifications
Specifications
CSA Class AL
Corded
Universal
Patented flange design for most comfortable fit of any reusable plug
26 dB
Yellow
Ultrafit Plus™ Earplug
No
Elastomeric Polymer
Multi-flanged
SDS