missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ E•A•R™ Push-Ins™ SofTouch Ear Plugs

NRR: 31dB

$75.73 - $119.08

Specifications

Certifications/Compliance CSA Class AL
Color Yellow
Product Type Push-Ins™ Softouch Earplug
Size One size fits to all
For Use With (Application) Tapered-cone shape and smooth, soft foam provide in-ear comfort and hearing protection
View More Specs

Alert:

The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.



Products ${productFamilyLength}
Catalog Number Mfr. No. Style Price Quantity  
Catalog Number Mfr. No. Style Price Quantity  
18-120-253 3M™
3184000
Uncorded Pack of 200 Each for $75.73
Case of 10 Pack for $688.40
18-120-254 3M™
3184001
Corded Pack of 200 Each for $119.08
Case of 10 Pack for $1,082.50
Specifications

Description

  • Specially formulated no-roll foam
  • Pistol-grip handle eliminates dirt transfer
  • Tapered cones for easy fit and comfort
  • Yellow with blue cord or uncorded

Description & Specifications

Specifications

CSA Class AL
Push-Ins™ Softouch Earplug
Tapered-cone shape and smooth, soft foam provide in-ear comfort and hearing protection
31dB
Yellow
One size fits to all
Polyurethane
Push-to-fit Foam