Please login to your online account to display your discounted pricing

3M™ Disposable Polypropylene Protective Lab Coat 4400

$198.10 - $206.40

Specifications

Closure Type Zipper front
Color Blue
Construction Breathable SMS, Microporous Laminate
Cuff Style Knit
Material Polypropylene, SMS
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

17-624-121

 
3m canada inc
44003XL
3XL Case of 50 Each for $206.40

17-624-122

 
3m™
44004XL
4XL Case of 50 Each for $198.10
Description & Specifications

Specifications

Closure Type Zipper front
Color Blue
Construction Breathable SMS, Microporous Laminate
Cuff Style Knit
Material Polypropylene, SMS
No. of Pockets 1.0
Disposable Yes
For Use With (Application) General maintenance, catering, electronics manufacturing, clean rooms, beauty industries, hygiene, laboratories
Recommended Storage -20 to 25°C
Seam Type Serged