Learn More
3M™ Comfort Plus 100 Class Disposable Filtering Facepiece Respirators
Provides a minimum filter efficiency of 99.97%
$13.05 - $318.36
Alert:
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog Number | Mfr. No. | Certifications/Compliance | Model | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Model | Price | Quantity | ||||
19-002711
|
3M™
7000002028 |
NIOSH 42 CFR Part 84, N100, NIOSH TC-84A-1298 | 8233 |
Each for $13.05
Case of 20 Each for $232.40 |
|
19-037352
|
3M™
7000002055 |
NIOSH 42 CFR Part 84, P100, NIOSH TC-84A-2561 | 8293 |
Case of 20 Each for $318.36
|
|
Description
- Ideal for certain OSHA substance specific contaminants (excluding asbestos but including lead, MDA, arsenic, and cadmium); additionally for welding and pharmaceutical manufacturing
- Provides NIOSH's highest-rated filtration efficiency in a disposable respirator
- Proprietary filter media with electrostatically charged microfibers designed for ease of breathing
- Cool Flow™ exhalation valve for easy exhalation and cool comfort
- Soft and secure faceseal conforms closely to face for additional comfort
- Proprietary P-Series Filter (Advanced Electrostatic Media) appropriate for prolonged use in both oil and non-oil containing environments (8293 only)
Adjustable buckle straps - Welding Web has unique filter media incorporating protective layers designed to be flame resistant as demonstrated per ASTM D2859-96 (not a substitute for a faceshield)
- 3M Catalog# 8233 , 3M Catalog# 8293
Compliance
Meets NIOSH 42 CFR 84 N100 requirements. NIOSH Approval No.: TC-84A-1298.