3M™ ClearVisor with Adflo™ PAPR System
Designed for non-welding applications
$1,644.14 - $1,644.14
Specifications
Airflow Rate 6+cfm
Certifications NIOSH 42CFR84, ANSI Z87.1-2003
Material Polycarbonate
Mounting Type Belt
View More Specs
Includes: ClearVisor faceshield assembly, filter, blower, filter cover, spark arrestor, prefilter, high-efficiency particulate filter, battery, smart charger, belt, breathing tube assembly and flow indicator. Select systems also include OV/SD/CL/HC cartridge.
ClearVisor shielding film available; see cat. no. 191500661.
Products ${productFamilyLength}
Catalog Number Mfr. No. Battery Life Product Type Price Quantity  
Catalog Number Mfr. No. Battery Life Product Type Price Quantity  
19-160-0338 3M™
16110140
12 to 13 Powered Air Purifying Respirator System Case for $1,644.14
Specifications
Description
Combines eye and face protection with a slim, lightweight powered air-purifying respirator. Chemical cartridge can be stacked onto the high-efficiency (HE) particulate filter for additional protection against organic vapors, sulfur dioxide, chlorine and hydrogen chloride (OV/SD/CL/HC).
ClearVisor Faceshield Assemby
  • For use in a variety of industrial settings, including woodworking, metal grinding, injection molding, foundries, pulp and paper processing and other dusty environments
  • Clear polycarbonate faceshield offers eye and face protection, exceptional clarity, and a large field of view
  • Attaches to fully adjustable, ratcheted headband with Velcro™ fasteners
  • Headband assembly features air duct , brow guard, cloth face seal, and cloth head cover
  • Continuous use of optional Clear Window Strip-Off Shielding Film extends useful service life of face shield (sold separately)
Adflo Powered Air-Purifying Respirator
  • Slim profile designed for tight spaces—no external batteries, battery cables or sharp corners
  • Brushless motor lasts three times longer than traditional motors
  • Solid leather belt is cut narrow in front for easy, comfortable forward bending: wider sides have large ventilation holes for maximum breathability
  • Turbo unit attaches securely between two arms of the belt to evenly distribute weight across the lower back and upper hips
  • Constant airflow rate regardless of battery charge and ilter loading; audible and visual alarms signal low airflow and low battery charge
  • NiMH battery charges in less than 3 hours with included smart charger and never overcharges
  • Comes standard with HE particle filter; also available with added cartridge for protection against OV/SD/CL/HC vapors
  • Filter and cartridge can be selectively replaced as needed—no need to change together
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Description & Specifications
Specifications
6+cfm
NIOSH 42CFR84, ANSI Z87.1-2003
Polycarbonate
Belt