3M™ Canister FR-40 and Cartridge FR-57
Tested to military standards
Manufacturer:  3M™ 70070889665
 View more versions of this product
Catalog No. 29-635-6648
Description
Description
  • Filter media use advanced 3M carbon filtering technology to protect the user from a wide range of contaminants
  • Filters a variety of chemical warfare agents including certain nerve agents, mustard agents, tear agents, blood agents, phosgene, chloropicrin and diphenylchloroarsine
  • 3M Catalog# 453-03-02R06
Manufacturer advises that spent cartridges and canisters be disposed in accordance with applicable federal, state and local laws and regulations. These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Certifications
NIOSH-approved for organic vapor, acid gas (sulfur dioxide, chlorine and hydrogen chloride), ammonia, phosphine, CS, CN and high efficiency filter.
Specifications
Specifications
Multi-gas and Vapor Cartridge
Black
6/Pk.
Rrpas 6000 series facepiece, be-10 series hood
NIOSH
453-03-02R06
High efficiency particulate protection in environments containing toxic industrial chemicals
Fr-57 Respirator Cartridge