3M™ BX™ Readers Protective Eyewear
Dual diopter available
Manufacturer:  3M™ 7000052801
 View more versions of this product
Catalog No. 19-065-403
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
  • High-impact polycarbonate lens absorbs 99.9% UV
  • Adjustable soft-tip temples feel comfortable even after extended wear
  • Soft nose bridge
  • Three lens angles for custom fit
  • 3M Catalog# 11374-00000-20, 11375-00000-20, 11376-00000-20, 11377-00000-20, 11378-00000-20, 11379-00000-20, 11457-00000-20, 11458-00000-20, 11475-00000-20, 11473-00000-20, 11474-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Specifications
ANSI Z87.1-2015
+2.00
Indoor/Outdoor Mirror
Reader Protective Eyewear
Ocean Blue
99.90%
Scratch-resistant
Documents