3M™ BX™ Readers Protective Eyewear
Dual diopter available
Manufacturer:  3M™ 113760000020
 View more versions of this product
Catalog No. 19-130-4584
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
  • High-impact polycarbonate lens absorbs 99.9% UV
  • Adjustable soft-tip temples feel comfortable even after extended wear
  • Soft nose bridge
  • Three lens angles for custom fit
Compliance
All models meet ANSI™ Z87.1-2010. Single diopter models also meet CSA Z94.3-2007 standards.
Specifications
Specifications
ANSI Z87.1-2010
+2.50
99.90%
Scratch-resistant
Silver
Reader Protective Eyewear
Clear
Documents