3M™ Breathe Easy™ Belt-Mounted Turbo PAPR Assemblies
Choose from two styles
$940.23 - $940.23
Specifications
Product Type | Powered Air Purifying Respirator Assembly |
---|---|
Filter Type | HE Filter |
Includes | Motor blower, belt, airflow indicator, battery pack |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
29-632-480
|
3M™
70070889285 |
450-00-01r12 he (p3) filter, intrinsically safe |
Each for $940.23
|
|
Description
Each assembly includes motor blower, belt, airflow indicator, and battery pack. For use with appropriate 3M filter, filter/cartridge or canister, and respirator headpiece.
3M Breathe Easy Belt-Mounted PAPRs
To assemble a complete system, select from the following pieces:
- Headgear, facepiece, head cover or hood
- Compatible breathing tube
- Breathe Easy belt-mounted Turbo PAPR assembly
- Appropriate cartridge/filter pack for your application
- Battery pack and charger
- 3M Catalog# 520-17-00
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product.
Specifications
Powered Air Purifying Respirator Assembly | |
HE Filter | |
Motor blower, belt, airflow indicator, battery pack |