3M™ Belt-Mounted and Vehicle-Mounted PAPR System Accessory, Cartridges
Choose from a large selection of cartridges for powered air purifying respirators used in a variety of industrial environments
Manufacturer: 3M™ GVP441
Description
- Cartridge attaches to the motor blower
- Filters air supplied to the headgear
- 3M Catalog# GVP-440, GVP-441, GVP-443, GVP-444, GVP-445
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product.
Specifications
Organic Vapor, High Efficiency (HE) | |
Gray Cartridge (Magenta/Black Label) | |
Transportation, Mining, Oil & Gas, General Manufacturing | |
GVP-441 |
NIOSH | |
Cartridge | |
Belt-mounted and vehicle-mounted PAPR systems | |
6/Cs. |