Please login to your online account to display your discounted pricing

3M™ Advanced Particulate Filters

$698.27 - $698.27

Specifications

Certifications NIOSH
Color Magenta
Connection Type Bayonet
Product Type Advanced Particulate Filter
For Use With (Application) For environments containing certain oil and non-oil based particles
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W x H) Filter Type Price Quantity    

19-177-844

 
3m™
2296
4.3 x 9.3 x 4.3 in. (10.92 x 23.62 x 10.92cm) 2296 Series Case of 100 Each for $698.27
Description & Specifications

Specifications

Certifications NIOSH
Color Magenta
Connection Type Bayonet
Product Type Advanced Particulate Filter
For Use With (Application) For environments containing certain oil and non-oil based particles
Length (English) 4.3 in.
Quantity 2 Pack