missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Adflo™ PAPR Accessory, Prefilter
Replacement particulate prefilter for 3M™ Adflo™ PAPR. Installs into the turbo filter cover between the spark arrestor and the HE particulate filter.
$33.89 - $59.05
Specifications
Color | White |
---|---|
Product Type | Prefilter |
For Use With (Equipment) | Adflo™ PAPR Systems |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
19-150-0600
|
3M™
7000126732 |
12/Cs. |
Case of 12 Each for $59.05
|
|
|||||
19-150-0599
|
3M™
7000126731 |
6/Cs. |
Case of 6 Each for $33.89
|
|
|||||
Description
- Particulate Filter
- Replacement Part
- 3M Catalog# 15-0099-99X12
Specifications
White | |
Prefilter | |
Adflo™ PAPR Systems |