3M™ Accessories and Replacement parts for Powerflow™ PAPR Systems
Used for respiratory protection
$506.62 - $573.15
Specifications
For Use With (Equipment) Powerflow™ face mounted powered air purifying respirator system
View More Specs
Products 2
Catalog Number Mfr. No. Type Applications Price Quantity  
Catalog Number Mfr. No. Type Applications Price Quantity  
29-663-290
View Documents
3M™
7000126344
Motor Blower Assembly Ideal to deliver minimum of 4 cfm when used with required high efficiency filter
Each for $506.62
29-635-7476
View Documents
3M™
7000131701
Sp3 Hepa Filter Ideal to protect against dusts, mists, fumes, asbestos, radionuclides and radon daughters
Case of 20 Each for $573.15
Description
Description
  • 3M Catalog# 024-00-02R01 , 3M Catalog# 450-01-01R20
Note: These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Specifications
Powerflow™ face mounted powered air purifying respirator system