3M™ 7500 Series Reusable Half Face Mask Respirator
State-of-the-art features provide unsurpassed comfort. The 3M Reusable Half Face Mask Respirator uses a Bayonet Attachment System for twin lightweight filters to protect against gases, vapors, and particulates.
Manufacturer:  3M™ 7100134949
 View more versions of this product
Catalog No. 19-096-900
Requires : Compatible cartridges and/or filters. Accommodates 3M 7093 and Series 2000 filters and Series 6000 cartridges.
Description
Description
Durable, reusable and heat-resistant mask is made of advanced silicone material that feels softer on the face, while the unique adjustment design helps reduce tension and pressure points on the face. 
  • Cool Flow™ exhalation valve opens easier, lowering exhalation breathing resistance, reducing heat and moisture buildup in the facepiece
  • Easily adjusted, dual-mode head harness gives user the option of wearing the respirator in either the traditional or drop-down mode
  • Yoke won't get hot like metal yokes, so the 7500 is an excellent choice for welding applications
  • Color-coded sizes for easy identification
  • 3M Catalog# 7501 , 3M Catalog# 7502 , 3M Catalog# 7503
 These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Compliance
NIOSH approved under 42 CFR 84 standard for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters; in Canada, CSA standard Z94.4 and applicable local government requirements must be met for approval.
Specifications
Specifications
Silicone
Bayonet
Large
Cartridge 6000 series, filter 2000, 2200, 7000 or 5000 series, or dual airline system
NIOSH 42 CFR Part 84
Half Mask Respirator
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
7503
SDS
Documents
Product Certifications
Accessories