Please login to your online account to display your discounted pricing

3M™ 4565 Series Disposable Protective Coveralls

$314.95 - $343.86

Specifications

Ankle Style Elastic
Certifications/Compliance ASTM F1670, EN 14126:2003, EN 1149-1:2006, EN 13034, EN-ISO 13982-30
Coating Anti-static
Closure Type Two-way Zipper with Sealed Storm Flap
Color White
View More Specs
Products
Catalog Number Mfr. No. Height (English) Height (Metric) Size Price Quantity    

17-623-364

 
3m™
45654XL
6.5 to 6.75 ft. 200 to 206cm 4X-Large Case of 25 Each for $314.95

17-623-359

 
3m™
4565M
5.5 to 5.75 ft. 167 to 176cm Medium Case of 20 Each for $343.86

17-623-360

 
3m™
4565L
5.75 to 6 ft. 174 to 181cm Large Case of 20 Each for $343.86

17-623-362

 
3m™
4565XXL
6 to 6.33 ft. 186 to 194cm 2X-Large Case of 20 Each for $343.86

17-623-361

 
3m™
4565XL
5.83 to 6.16 ft. 179 to 187cm X-Large Case of 20 Each for $343.86

17-623-363

 
3m™
45653XL
6.33 to 6.5 ft. 194 to 200cm 3X-Large Case of 25 Each for $314.95
Description & Specifications

Specifications

Ankle Style Elastic
Certifications/Compliance ASTM F1670, EN 14126:2003, EN 1149-1:2006, EN 13034, EN-ISO 13982-30
Coating Anti-static
Closure Type Two-way Zipper with Sealed Storm Flap
Color White
Construction Polyethylene and Polyester/Polyethylene laminate suit, Metal/Nylon/Polyester Braid zipper, Neoprene Rubber elastic, Polyester/Cotton thread, Polyester cuffs, Polypropylene red tape
Cuff Style Elastic
Material Microporous Polyethylene
Seam Type Taped
Disposable Yes
For Use With (Application) Paint spraying, industrial and pharmaceutical manufacturing, asbestos inspection and handling and healthcare