missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ White Protective Coverall 4520
Protective Coveralls that provide protection against liquid splashes and dust.
Supplier: 3M™ 7000089643
Description
- Designed to offer protection against hazardous dusts and limited liquid splashes and sprays
- Made from lightweight and breathable material for reduced heat stress and premium wearer comfort all day
- Standard collar for use with respiratory equipment or where a hood may not be desired
- Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
Specifications
3X-Large | |
White | |
Elastic, Knit | |
Bound | |
CE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006 | |
4520-3 XL | |
35 in. | |
124.46 to 132.08 cm | |
137.16 cm | |
6.33 to 6.5 ft. |
Polypropylene | |
Single-use | |
2-way Zipper with Storm Flap | |
Anti-static | |
White SMMMS Polypropylene suit and green back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread | |
49 to 52 in. | |
54 in. | |
88.9 cm | |
Insulation Installation, Asbestos and Lead Abatement, Coal Dust in Power Plants, Woodworking, Metal Polishing, Light-Duty Building Cleaning, General Powder Handling, General Industrial Clean-up and Maintenance, Machine or Vehicle Maintenance and Food Processing | |
194 to 200 cm |