missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Hooded Protective Coveralls 4510 Series, Elastic Wrist and Ankles
Coveralls provide light duty protection against infective agents, certain hazardous dusts, and limited liquid splashes. PE laminate material includes micropores that allow heat and moisture to evaporate from the skin and out through the suit for a lightweight and comfortable fit.
Supplier: 3M™ 7000089665
Description
- Type 5/6 Coverall designed to offer cost effective protection against hazardous dusts and limited liquid splashes
- Protection against EN 14126 Biological agents
- Made of a soft lightweight non-woven with microporous laminate for splash protection, strength and breathability
- Certified protection against Type 5/6 hazardous dusts and limited liquid splashes
- 2 panel hood design, 2-way zip with storm flap, elasticated apertures reduce the risk of skin exposure
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
Specifications
Cleanroom Coveralls | |
Microporous Polyethylene Laminate | |
Single-use | |
Elastic | |
Bound | |
Anti-static | |
White Polypropylene/Polyethylene laminate film suit, Nylon on Polyester Braid zipper, Neoprene Rubber elastic, Polyester thread | |
4510-XXL | |
34 in. | |
114.3 to 124.46 cm | |
134.62 cm | |
6 to 6.33 ft. |
2X-Large | |
White | |
Elastic | |
2-way Zipper with Storm Flap | |
Non-sterile | |
EN 13034 Type 6, EN ISO 13982-1 Type 5, EN 1149-1:2006, ASTM D2859-96, CE | |
Yes | |
45 to 49 in. | |
53 in. | |
86.36 cm | |
Abatement, Remediation, Agriculture, Cleaning and Maintenance, General Industry, Manufacturing, Non-Hazardous Operations, Painting, Pharmaceutical Cleanroom | |
186 to 194 cm |