missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ 1000 Ear Muff

$18.27 - $1,122.29

Specifications

Size One size fits most
Material ABS
Noise Reduction Rating (NRR) 20dB over-the-head and 22dB behind-the-neck
View More Specs

Products 2
Catalog Number Mfr. No. Color Product Type Style Price Quantity  
Catalog Number Mfr. No. Color Product Type Style Price Quantity  
17-380-10 View Documents 3M™
3303001
Black/maroon Ear Muff Multiposition Each for $18.27
Case of 24 Each for $402.96
17-381A View Documents 3M™
3313010
Black Ear Muff Cushion Over-the-head, behind-the-neck Case of 150 Pair for $1,122.29
Description

Description

NRR: 20dB over-the-head; NRR: 22dB behind-the-neck.

Made from lightweight, durable, nonconductive plastic. Pivoting ear cup connection provides comfortable seal while maintaining alignment with head. Large ear cup openings are filled with foam cushions. Take-apart assembly makes cleaning easy. Adjustable headband lets workers choose from three positions: over-the-head Noise Reduction Rating (NRR): 20dB, under-the-chin NRR: 22dB, and behind-the-neck NRR: 22dB.

Note:When worn under the chin or behind the neck, an optional crown strap should be worn. To order, call your Fisher Safety Customer Service Representative at 1-800-772-6733.

Order Info

Net weight: 7 3/16 oz. (204g).

Specifications

Specifications

One size fits most
ABS
20dB over-the-head and 22dB behind-the-neck