missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3H-1,2-Benzodithiol-one 1,1-dioxide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L2012906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3H-1,2-Benzodithiol-one 1,1-dioxide | |
102°C to 107°C | |
MFCD00132960 | |
UN3335 | |
3h-1,2-benzodithiol-3-one 1,1-dioxide, beaucage reagent, 3h-1,2-benzodithiol-3-one-1,1-dioxide, 3h-benzo c 1,2 dithiol-3-one 1,1-dioxide, 3h-1,2-benzodithiol-3-one, 1,1-dioxide, 1,1-dioxo-1$l^ 6 ,2-benzodithiol-3-one, 3h-1,2-benzodithiol-one 1,1-dioxide, 3h-1,2-benzodithiol-3-one1,1-dioxide, 1??,2-benzodithiole-1,1,3-trione, acmc-1b2sk | |
C1=CC=C2C(=C1)C(=O)SS2(=O)=O | |
200.226 | |
200.24 |
66304-01-6 | |
C7H4O3S2 | |
5 g | |
4180332 | |
JUDOLRSMWHVKGX-UHFFFAOYSA-N | |
1,1-dioxo-1$l^{6},2-benzodithiol-3-one | |
3009847 | |
99% |
Chemical Identifiers
66304-01-6 | |
200.226 | |
JUDOLRSMWHVKGX-UHFFFAOYSA-N | |
1,1-dioxo-1$l^{6},2-benzodithiol-3-one |
C7H4O3S2 | |
MFCD00132960 | |
3009847 | |
C1=CC=C2C(=C1)C(=O)SS2(=O)=O |
Safety and Handling
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only