Please login to your online account to display your discounted pricing

Alfa Aesar™ 3H-1,2-Benzodithiol-one 1,1-dioxide, 99%


Quantity 1g

Manufacturer: alfa aesar™  L2012903

 View more versions of this product

Catalog No. AAL2012903

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 200.24
Chemical Name or Material 3H-1,2-Benzodithiol-one 1,1-dioxide
MDL Number MFCD00132960
Assay 99%
Molecular Formula C7H4O3S2
CAS 66304-01-6
UN Number UN3335
Beilstein 4180332
Synonym Beaucage Reagent
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
Melting Point 102° to 107°C
Quantity 1g

Specifications

Chemical Name or Material 3H-1,2-Benzodithiol-one 1,1-dioxide
CAS 66304-01-6
Molecular Formula C7H4O3S2
Formula Weight 200.24
MDL Number MFCD00132960
UN Number UN3335
Beilstein 4180332
Synonym Beaucage Reagent