Please login to your online account to display your discounted pricing

3H-1,2-benzodithiol-3-one 1,1-dioxide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3H-1, 2-Benzodithiol-3-one 1, 1-dioxide
CAS 66304-01-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H4 O3 S2
Formula Weight 200.24
MDL Number MFCD00132960
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379340010

  SDS
thermo fisher scientific
AC379340010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC379342500

  SDS
thermo fisher scientific
AC379342500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystals
Formula Weight 200.24
Color White tranparent
Quantity 1g
Melting Point 102° to 107°C
CAS Max % 100.0
Chemical Name or Material 3H-1, 2-Benzodithiol-3-one 1, 1-dioxide
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C7 H4 O3 S2
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
MDL Number MFCD00132960
CAS 66304-01-6
CAS Min % 97.5
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.