missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3H-1,2,3-Triazolo[4,5-b]pyridin-3-ol, TCI America™

Manufacturer:  TCI America T16735G

 View more versions of this product

Catalog No. T16735G



Specifications

Specifications

3H-1,2,3-Triazolo[4,5-b]pyridin-3-ol
5g
C5H4N4O
0481
FPIRBHDGWMWJEP-UHFFFAOYSA-N
3-hydroxytriazolo[4,5-b]pyridine
181649
Crystal-Powder at 20°C
White-Yellow
39968-33-7
MFCD00210053
1-hydroxy-7-azabenzotriazole, hoat, 3h-1,2,3 triazolo 4,5-b pyridin-3-ol, 3-hydroxytriazolo 4,5-b pyridine, unii-tx8xyh09h0, 1-hydroxy-7-azabenzotriazole hoat, hoat 1-hydroxy-7-azabenzotriazole, 3h-1,2,3-triazolo 4,5-b pyridine, 3-hydroxy, tx8xyh09h0, 7-aza-1-hydroxybenzotriazole
C1=CC2=C(N=C1)N(N=N2)O
136.114
136.11
≥98.0% (HPLC,T)

Chemical Identifiers

39968-33-7
136.114
FPIRBHDGWMWJEP-UHFFFAOYSA-N
181649
C1=CC2=C(N=C1)N(N=N2)O
C5H4N4O
MFCD00210053
1-hydroxy-7-azabenzotriazole, hoat, 3h-1,2,3 triazolo 4,5-b pyridin-3-ol, 3-hydroxytriazolo 4,5-b pyridine, unii-tx8xyh09h0, 1-hydroxy-7-azabenzotriazole hoat, hoat 1-hydroxy-7-azabenzotriazole, 3h-1,2,3-triazolo 4,5-b pyridine, 3-hydroxy, tx8xyh09h0, 7-aza-1-hydroxybenzotriazole
3-hydroxytriazolo[4,5-b]pyridine
Safety and Handling

Safety and Handling

TSCA : No

Documents