Please login to your online account to display your discounted pricing

3F8, Tocris™

Potent and selective GSK-3β inhibitor

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material 3F8
Assay Percent Range >99%
Molecular Formula C15H14N2O4
Formula Weight 286.28
Synonym 5-Ethyl-7,8-dimethoxy-1H-pyrrolo[3,4-c]isoquinoline-1,3(2H)-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

408310

 
r & d systems
408310
10mg Each for $225.77

408350

 
r & d systems
408350
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 286.28
Quantity 10mg
Chemical Name or Material 3F8
Assay Percent Range >99%
Molecular Formula C15H14N2O4
Synonym 5-Ethyl-7,8-dimethoxy-1H-pyrrolo[3,4-c]isoquinoline-1,3(2H)-dione
CAS 159109-11-2
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C