3B Scientific™ Low Form Beakers
Low form beakers are made up of transparent TPX material.
$16.15 - $16.15
Specifications
Color Transparent
Material PMP (Polymethylpentene)
Product Type Beaker
Graduated No
Height (English) 2.75 in.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S29331 3B Scientific™
U29331
200mL Each for $16.15
Dozen of 12 Each for $193.60
Specifications Description & Specifications
Specifications
Transparent
Beaker
2.75 in.
PMP (Polymethylpentene)
No
7cm