missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3B Scientific™ Graduated Cylinders

Made up of transparent TPX material.

Manufacturer:  3B Scientific™ U29332

 View more versions of this product

Catalog No. S29332



 

Specifications

Description

Cylinder; 3B Scientific; Graduated; Material: Transparent TPX; Divisions: 1mL; Capacity: 50mL

  • Graduated (1mL divisions)
  • Material: Transparent TPX
Description & Specifications

Specifications

50mL
1mL
1mL
PMP (Polymethylpentene)
Yes