Please login to your online account to display your discounted pricing

3B Scientific™ Fetus Skull


Study the anatomy of a human fetal skull with this realistic replica.

Manufacturer: 3b scientific™  A26

 View more versions of this product

Catalog No. S17135T

  • / Each
 

Description & Specifications

Specifications

Type Anatomical and cellular Human fetus skull model
Mounting Stand
Weight (English) 0.44 lb.
Material Plastic
Includes Stand
Weight (Metric) 0.2kg