missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3B Scientific™ Brain Models

Different separations of the brain expose different levels of detail and complexity.

Manufacturer:  3B Scientific™ C15

 View more versions of this product

Catalog No. S17108



Description

Description

  • Vinyl plastic models are nonbreakable and natural cast
  • Rest on a form-fitting molded base
  • Dimensions: 5 × 6 × 5 in. (12.7 × 15.2 × 12.7cm)

Two-Part Brain

  • Median section divides brain into two sections
  • Four-Part Brain
    • Median section dissectible into two vertical halves
    • Right half is dissectible into brain lobe and brain stem with occipital lobe

    Eight-Part Brain
    • Each median section is dissectible into frontal with parietal lobes, temporal with occipital lobes, half of cerebellum and half of brain stem
    • Superb coloration highlights variety of brain systems

    Specifications

    Specifications

    For beginning studies
    Vinyl
    Anatomical and cellular Human brain model
    0.7kg
    Form fitting molded base
    Molded Base
    1.54 lb.
    Documents