missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3B Scientific™ Anatomy Posters
Visual aids to enhance anatomy lessons.
Manufacturer: 3B Scientific™ VR1334L
Description
- Printed on premium glossy (200g) paper
- With two-sided lamination (125 micron) and metal eyelets
- Dimensions: 20W × 26 in.H (50.8 × 66.0cm)
Specifications
The Human Heart - Anatomy and Physiology Chart | |
26 in. | |
20 in. | |
Great teaching aid for lessons on the Human heart and the Human circulatory system |
Science | |
66cm | |
50.8cm |