missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3B Scientific™ Anatomy Posters

Visual aids to enhance anatomy lessons.

Manufacturer:  3B Scientific™ VR1322L

 View more versions of this product

Catalog No. S90763



Description

Description

  • Printed on premium glossy (200g) paper
  • With two-sided lamination (125 micron) and metal eyelets
  • Dimensions: 20W × 26 in.H (50.8 × 66.0cm)

Specifications

Specifications

The Respiratory System Chart
26 in.
20 in.
Important part of respiratory anatomy is pictured and labeled including the respiratory musculature, the bronchial tree and the lungs
Science
66cm
50.8cm
SDS
Documents
Product Certifications
Promotions

Promotions