missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3aR,4S,5R,6aS)-(-)-Hexahydro-5-hydroxy-4-hydroxymethyl-2H-cyclopenta[b]furan-2-one, 98%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H56119MD

 View more versions of this product

Catalog No. AAH56119MD



Description

Description

Specifications

Specifications

(3aR,4S,5R,6aS)-(-)-Hexahydro-5-hydroxy-4-hydroxymethyl-2H-cyclopenta[b]furan-2-one
117°C to 119°C
32233-40-2
MFCD00075234
VYTZWRCSPHQSFX-GBNDHIKLSA-N
(3aR,4S,5R,6aS)-5-hydroxy-4-(hydroxymethyl)-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-2-one
2724453
98%
−44° (c=1.4 in Methanol)
250mg
C8H12O4
--corey diol corey lactone, --corey lactone, --corey lactone diol, 1s,6s,5r,7r-7-hydroxy-6-hydroxymethyl-2-oxabicyclo 3.3.0 octan-3-one, 3ar,4s,5r,6as---hexahydro-5-hydroxy-4-hydroxymethyl-2h-cyclopenta b furan-2-one, 3ar,4s,5r,6as-5-hydroxy-4-hydroxymethyl hexahydro-2h-cyclopenta b furan-2-one, 3ar,4s,5r,6as-5-hydroxy-4-hydroxymethyl-3,3a,4,5,6,6a-hexahydrocyclopenta b furan-2-one, 3ar,4s,5r,6as-hexahydro-5-hydroxy-4-hydroxymethyl-2h-cyclopenta b furan-2-one, corey lactone diol, l-corey lactone
OC[C@H]1[C@H](O)C[C@@H]2OC(=O)C[C@H]12
172.18
172.18

Chemical Identifiers

32233-40-2
172.18
VYTZWRCSPHQSFX-GBNDHIKLSA-N
2724453
OC[C@H]1[C@H](O)C[C@@H]2OC(=O)C[C@H]12
C8H12O4
MFCD00075234
--corey diol corey lactone, --corey lactone, --corey lactone diol, 1s,6s,5r,7r-7-hydroxy-6-hydroxymethyl-2-oxabicyclo 3.3.0 octan-3-one, 3ar,4s,5r,6as---hexahydro-5-hydroxy-4-hydroxymethyl-2h-cyclopenta b furan-2-one, 3ar,4s,5r,6as-5-hydroxy-4-hydroxymethyl hexahydro-2h-cyclopenta b furan-2-one, 3ar,4s,5r,6as-5-hydroxy-4-hydroxymethyl-3,3a,4,5,6,6a-hexahydrocyclopenta b furan-2-one, 3ar,4s,5r,6as-hexahydro-5-hydroxy-4-hydroxymethyl-2h-cyclopenta b furan-2-one, corey lactone diol, l-corey lactone
(3aR,4S,5R,6aS)-5-hydroxy-4-(hydroxymethyl)-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-2-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications