missing translation for 'onlineSavingsMsg'
Learn More

3a,4,7,7a-Tetrahydroindene (stabilized with BHT) 98.0+%, TCI America™

Supplier:  TCI America T1315500ML

 View more versions of this product

Catalog No. T1315500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3a,4,7,7a-Tetrahydroindene (stabilized with BHT)
Yellow
C9H12
500 mL
3a,4,7,7a-tetrahydroindene, 1h-indene, 3a,4,7,7a-tetrahydro, indene, 3a,4,7,7a-tetrahydro, 4,7,8,9-tetrahydroindene, bicyclo 4,3,0 nona-3,7-diene, tetrahydroindene russian, bicyclo 4.3.0 nona-3,7-diene, 1h-indene, 3a,4,7,7a-tetrahydro-, cis, cis-bicyclo 4.3.0 nona-3,7-diene, 1h-indene, 3a,4,7,7a-tetrahydro-, trans
C1C=CCC2C1CC=C2
120.195
120.20
Liquid
3048-65-5
160°C
MFCD00021227
3295
UFERIGCCDYCZLN-UHFFFAOYSA-N
3a,4,7,7a-tetrahydro-1H-indene
62475
≥98.0% (GC)

Chemical Identifiers

3048-65-5
120.195
UFERIGCCDYCZLN-UHFFFAOYSA-N
62475
C1C=CCC2C1CC=C2
C9H12
MFCD00021227
3a,4,7,7a-tetrahydroindene, 1h-indene, 3a,4,7,7a-tetrahydro, indene, 3a,4,7,7a-tetrahydro, 4,7,8,9-tetrahydroindene, bicyclo 4,3,0 nona-3,7-diene, tetrahydroindene russian, bicyclo 4.3.0 nona-3,7-diene, 1h-indene, 3a,4,7,7a-tetrahydro-, cis, cis-bicyclo 4.3.0 nona-3,7-diene, 1h-indene, 3a,4,7,7a-tetrahydro-, trans
3a,4,7,7a-tetrahydro-1H-indene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0581

RTECSNumber : NK8750000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions