Please login to your online account to display your discounted pricing

Alfa Aesar™ 3a,4,7,7a-Tetrahydroindene, 98%

Chemical Identifiers

Chemical Name or Material 3a,4,7,7a-Tetrahydroindene
CAS 3048-65-5
Molecular Formula C9H12
Formula Weight 120.2
MDL Number MFCD00021227
UN Number UN3295
Beilstein 1902859
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1040909

  SDS
alfa aesar™
L1040909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1040918

  SDS
alfa aesar™
L1040918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.2
RTECS Number NK8750000
Chemical Name or Material 3a,4,7,7a-Tetrahydroindene
Assay 98%
Health Hazard 2 GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Molecular Formula C9H12
MDL Number MFCD00021227
CAS 3048-65-5
UN Number UN3295
Beilstein 1902859
Health Hazard 1 Danger
Boiling Point 158° to 160°C
Flash Point 38°C (100°F)
Quantity 10g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.