missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3a,4,7,7a-Tetrahydroindene, 98%

Manufacturer:  Alfa Aesar™ L1040918

 View more versions of this product

Catalog No. AAL1040918



Specifications

Specifications

3a,4,7,7a-Tetrahydroindene
158°C to 160°C
3048-65-5
MFCD00021227
1902859
UFERIGCCDYCZLN-UHFFFAOYSA-N
3a,4,7,7a-tetrahydro-1H-indene
62475
98%
38°C (100°F)
50g
C9H12
UN3295
3a,4,7,7a-tetrahydroindene, 1h-indene, 3a,4,7,7a-tetrahydro, indene, 3a,4,7,7a-tetrahydro, 4,7,8,9-tetrahydroindene, bicyclo 4,3,0 nona-3,7-diene, tetrahydroindene russian, bicyclo 4.3.0 nona-3,7-diene, 1h-indene, 3a,4,7,7a-tetrahydro-, cis, cis-bicyclo 4.3.0 nona-3,7-diene, 1h-indene, 3a,4,7,7a-tetrahydro-, trans
C1C=CCC2C1CC=C2
120.195
120.2

Chemical Identifiers

3048-65-5
120.195
UFERIGCCDYCZLN-UHFFFAOYSA-N
62475
C1C=CCC2C1CC=C2
C9H12
MFCD00021227
3a,4,7,7a-tetrahydroindene, 1h-indene, 3a,4,7,7a-tetrahydro, indene, 3a,4,7,7a-tetrahydro, 4,7,8,9-tetrahydroindene, bicyclo 4,3,0 nona-3,7-diene, tetrahydroindene russian, bicyclo 4.3.0 nona-3,7-diene, 1h-indene, 3a,4,7,7a-tetrahydro-, cis, cis-bicyclo 4.3.0 nona-3,7-diene, 1h-indene, 3a,4,7,7a-tetrahydro-, trans
3a,4,7,7a-tetrahydro-1H-indene
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

RTECSNumber : NK8750000

TSCA : TSCA

Documents