Thermo Scientific™ 384-Well Full Skirted PCR Plates
Reduce well-to-well variability with 384-Well PCR Plate Full Skirted PCR Plates, thin walled for efficient heat transfer and compatible with most major thermal cyclers.
$664.05 - $664.05
Specifications
Certifications/Compliance | Certified free of human DNA, RNase, DNase, and endotoxins. |
---|---|
Format | Standard |
Array | 16 x 24 |
No. of Wells | 384 |
Volume (Metric) Working | 25 μL |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
TF-0384
|
Thermo Scientific™
TF0384 |
Natural |
Case of 50 Each for $664.05
|
|
Description
384-Well Full Skirted PCR Plates are thin walled for efficient heat transfer and compatible with most major thermal cyclers.
Highlights
- Raised rim design around each well enables secure sealing and safeguards against evaporation
- Uniform, thin well walls deliver maximum and consistent heat transfer for equally high performance from every sample
- SBS footprint
- 40μL maximum well volume, 25μL working volume
- Cut Corner: A24
Specifications
Certified free of human DNA, RNase, DNase, and endotoxins. | |
16 x 24 | |
25 μL |
Standard | |
384 |